Assay Details
| Gene Symbol: CCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102382.1 | NP_001095852.1 | 3 - 4 | 614 | 126 | |
| GenBank mRNA | BC113333.1 | - | 3 - 4 | 614 | 126 | |
More Information
Set Membership: |
|

CCN3
Genomic Map