Assay Details
| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102491.1 | NP_001095961.1 | 6 - 7 | 1470 | 63 | |
| GenBank mRNA | BC151392.1 | - | 6 - 7 | 1470 | 63 | |
More Information
Set Membership: |
|

FAM160A2
Genomic Map