Assay Details
| Gene Symbol: RAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102505.1 | NP_001095975.1 | 8 - 9 | 1259 | 63 | |
| GenBank mRNA | BC151319.1 | - | 8 - 9 | 1259 | 63 | |
More Information
Set Membership: |
|

RAF1
Genomic Map