Assay Details
| Gene Symbol: MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145793.1 | NP_001139265.1 | 6 - 7 | 767 | 129 | |
| GenBank mRNA | EW684280.1 | - | 3 - 4 | 665 | 129 | |
| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 10 - 11 | 1141 | 129 | ||
More Information
Set Membership: |
|

MEF2B
Genomic Map