Assay Details
| Gene Symbol: MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145793.1 | NP_001139265.1 | 8 - 9 | 967 | 130 | |
| GenBank mRNA | EW684280.1 | - | 1 - 2 | 465 | 130 | |
| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 12 - 13 | 1341 | 130 | ||
More Information
Set Membership: |
|

MEF2B
Genomic Map