Assay Details
| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 5 - 6 | 432 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100271850-MEF2B| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 5 - 6 | 432 | 106 | ||
Set Membership: |
|