Assay Details
| Gene Symbol: CHRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103243.1 | NP_001096713.1 | 1 | 2054 | 65 | |
| GenBank mRNA | BC149788.1 | - | 1 | 2054 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRM4| Gene Symbol: CHRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103243.1 | NP_001096713.1 | 1 | 2054 | 65 | |
| GenBank mRNA | BC149788.1 | - | 1 | 2054 | 65 | |
Set Membership: |
|