Assay Details
| Gene Symbol: CYB5R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103250.1 | NP_001096720.1 | 5 - 6 | 487 | 68 | |
| GenBank mRNA | BC146232.1 | - | 5 - 6 | 487 | 68 | |
| M83104.1 | - | 4 - 5 | 460 | 68 | ||
More Information
Set Membership: |
|

CYB5R3
Genomic Map