Assay Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104965.1 | NP_001098435.1 | 1 - 2 | 109 | 139 | |
GenBank mRNA | BC149662.1 | - | 1 - 2 | 109 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149662.1, NM_001104965.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
