Assay Details
| Gene Symbol: ZNF266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104968.1 | NP_001098438.1 | 3 - 4 | 288 | 69 | |
| GenBank mRNA | BC149066.1 | - | 3 - 4 | 288 | 69 | |
More Information
Set Membership: |
|

ZNF266
Genomic Map