Assay Details
| Gene Symbol: MOGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104970.1 | NP_001098440.1 | 1 - 2 | 124 | 134 | |
| GenBank mRNA | BC149059.1 | - | 1 - 2 | 124 | 134 | |
More Information
Set Membership: |
|

MOGAT2
Genomic Map