Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104971.1 | NP_001098441.1 | 3 - 4 | 616 | 85 | |
| GenBank mRNA | BC142476.1 | - | 3 - 4 | 616 | 85 | |
More Information
Set Membership: |
|

PAIP1
Genomic Map