Assay Details
| Gene Symbol: ASPSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104980.1 | NP_001098450.1 | 11 - 12 | 1310 | 57 | |
| GenBank mRNA | BC142388.1 | - | 11 - 12 | 1310 | 57 | |
More Information
Set Membership: |
|

ASPSCR1
Genomic Map