Assay Details
| Gene Symbol: GPM6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104981.1 | NP_001098451.1 | 3 - 4 | 500 | 58 | |
| GenBank mRNA | BC151438.1 | - | 3 - 4 | 500 | 58 | |
More Information
Set Membership: |
|

GPM6B
Genomic Map