Assay Details
| Gene Symbol: INO80B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105347.1 | NP_001098817.1 | 3 - 4 | 377 | 120 | |
| GenBank mRNA | BC153273.1 | - | 3 - 4 | 377 | 120 | |
More Information
Set Membership: |
|

INO80B
Genomic Map