Assay Details
| Gene Symbol: MED19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105348.1 | NP_001098818.1 | 1 - 2 | 250 | 80 | |
| GenBank mRNA | BC134635.1 | - | 1 - 2 | 250 | 80 | |
More Information
Set Membership: |
|

MED19
Genomic Map