Assay Details
| Gene Symbol: CPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105350.2 | NP_001098820.1 | 10 - 11 | 1079 | 66 | |
| GenBank mRNA | BC134793.1 | - | 10 - 11 | 1080 | 66 | |
| EU515132.1 | - | 10 - 11 | 1068 | 66 | ||
More Information
Set Membership: |
|

CPB1
Genomic Map