Assay Details
| Gene Symbol: FITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105351.2 | NP_001098821.1 | 1 - 2 | 782 | 72 | |
| GenBank mRNA | BC134663.1 | - | 1 - 2 | 782 | 72 | |
More Information
Set Membership: |
|

FITM1
Genomic Map