Assay Details
| Gene Symbol: HDGFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105361.1 | NP_001098831.1 | 13 - 14 | 1630 | 65 | |
| GenBank mRNA | BC153286.1 | - | 13 - 14 | 1630 | 65 | |
More Information
Set Membership: |
|

HDGFL2
Genomic Map