Assay Details
| Gene Symbol: HDGFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105361.1 | NP_001098831.1 | 1 - 2 | 133 | 126 | |
| GenBank mRNA | BC153286.1 | - | 1 - 2 | 133 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153286.1, NM_001105361.1 transcripts.
Set Membership: |
|

HDGFL2
Genomic Map