Assay Details
| Gene Symbol: HDGFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105361.1 | NP_001098831.1 | 4 - 5 | 542 | 82 | |
| GenBank mRNA | BC153286.1 | - | 4 - 5 | 542 | 82 | |
More Information
Set Membership: |
|

HDGFL2
Genomic Map