Assay Details
| Gene Symbol: COMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105368.2 | NP_001098838.1 | 1 - 2 | 211 | 63 | |
| GenBank mRNA | BC134766.1 | - | 1 - 2 | 212 | 63 | |
More Information
Set Membership: |
|

COMTD1
Genomic Map