Assay Details
| Gene Symbol: POLR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105622.2 | NP_001099092.1 | 1 - 2 | 271 | 103 | |
| GenBank mRNA | BC134470.1 | - | 1 - 2 | 272 | 103 | |
More Information
Set Membership: |
|

POLR1F
Genomic Map