Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109790.1 | NP_001103260.1 | 2 - 3 | 303 | 73 | |
| GenBank mRNA | BC153831.1 | - | 2 - 3 | 303 | 73 | |
More Information
Set Membership: |
|

FAM168B
Genomic Map