Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109791.1 | NP_001103261.1 | 6 - 7 | 3134 | 104 | |
| GenBank mRNA | BC149807.1 | - | 6 - 7 | 3134 | 104 | |
More Information
Set Membership: |
|

CRYBG2
Genomic Map