Assay Details
Gene Symbol: CYP2C19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109792.2 | NP_001103262.1 | 2 - 3 | 347 | 78 | |
GenBank mRNA | BC153845.1 | - | 2 - 3 | 349 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
