Assay Details
| Gene Symbol: RBIS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110068.2 | NP_001103538.1 | 1 - 2 | 55 | 79 | |
| GenBank mRNA | BC108159.1 | - | 1 | 56 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108159.1, NM_001110068.2 transcripts.
Set Membership: |
|

RBIS
Genomic Map