Assay Details
| Gene Symbol: FBXL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110077.2 | NP_001103547.1 | 2 - 3 | 1112 | 80 | |
| GenBank mRNA | BC149287.1 | - | 2 - 3 | 1113 | 80 | |
More Information
Set Membership: |
|

FBXL15
Genomic Map