Assay Details
| Gene Symbol: CXCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113172.1 | NP_001106643.1 | 3 - 4 | 293 | 146 | |
| GenBank mRNA | EU276061.1 | - | 3 - 4 | 293 | 146 | |
More Information
Set Membership: |
|

CXCL9
Genomic Map