Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034759.2 | NP_001029931.2 | 4 - 5 | 653 | 80 | |
| GenBank mRNA | BC102886.1 | - | 4 - 5 | 371 | 80 | |
| BT021503.1 | - | 4 - 5 | 374 | 80 | ||
| DV841387.1 | - | 4 - 5 | 654 | 80 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map