Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038575.4 | NP_001033664.2 | 1 - 2 | 670 | 132 | |
| NM_001314067.1 | NP_001300996.1 | 1 - 2 | 670 | 132 | ||
| GenBank mRNA | BC109678.1 | - | 1 - 2 | 666 | 132 | |
More Information
Set Membership: |
|

MINPP1
Genomic Map