Assay Details
| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046368.2 | NP_001039833.2 | 11 - 12 | 942 | 61 | |
| GenBank mRNA | BC105446.1 | - | 11 - 12 | 978 | 61 | |
| DV883546.1 | - | 8 - 9 | 624 | 61 | ||
More Information
Set Membership: |
|

VPS35L
Genomic Map