Assay Details
| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046368.2 | NP_001039833.2 | 19 - 20 | 1642 | 84 | |
| GenBank mRNA | DV907223.1 | - | 4 - 5 | 331 | 84 | |
More Information
Set Membership: |
|

VPS35L
Genomic Map