Assay Details
| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046368.2 | NP_001039833.2 | 23 - 24 | 1943 | 146 | |
| GenBank mRNA | DV907223.1 | - | 8 - 9 | 632 | 146 | |
More Information
Set Membership: |
|

VPS35L
Genomic Map