Assay Details
| Gene Symbol: RHBDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046478.2 | NP_001039943.2 | 1 - 2 | 263 | 62 | |
| GenBank mRNA | BC112680.1 | - | 1 - 2 | 214 | 62 | |
| DV892216.1 | - | 1 - 2 | 263 | 62 | ||
More Information
Set Membership: |
|

RHBDD2
Genomic Map