Assay Details
| Gene Symbol: RHBDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046478.2 | NP_001039943.2 | 3 - 4 | 819 | 123 | |
| GenBank mRNA | CO890373.1 | - | 2 - 3 | 176 | 123 | |
| DV892216.1 | - | 3 - 4 | 819 | 123 | ||
More Information
Set Membership: |
|

RHBDD2
Genomic Map