Assay Details
| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076084.2 | NP_001069552.2 | 3 - 4 | 427 | 64 | |
| GenBank mRNA | BC122785.1 | - | 3 - 4 | 428 | 64 | |
More Information
Set Membership: |
|

SUN3
Genomic Map