Assay Details
| Gene Symbol: PRSS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC148914.1 | - | 2 - 3 | 298 | 70 | |
| BT025375.1 | - | 276 | 70 | |||
| DT721783.1 | - | 2 - 3 | 296 | 70 | ||
| DV805457.1 | - | 2 - 3 | 296 | 70 | ||
| DY068401.1 | - | 2 - 3 | 278 | 70 | ||
| DY182405.1 | - | 2 - 3 | 289 | 70 | ||
More Information
Set Membership: |
|

PRSS16
Genomic Map