Assay Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076426.2 | NP_001069894.2 | 4 - 5 | 408 | 60 | |
GenBank mRNA | BC102597.1 | - | 4 - 5 | 409 | 60 | |
DT841167.1 | - | 4 - 5 | 408 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
