Assay Details
| Gene Symbol: PIGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077022.2 | NP_001070490.1 | 4 - 5 | 371 | 69 | |
| GenBank mRNA | BC108167.1 | - | 4 - 5 | 366 | 69 | |
| BM430006.1 | - | 1 - 2 | 213 | 69 | ||
| DY460750.1 | - | 4 - 5 | 375 | 69 | ||
More Information
Set Membership: |
|

PIGP
Genomic Map