Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080297.2 | NP_001073766.2 | 9 - 10 | 1449 | 58 | |
| GenBank mRNA | BC146071.1 | - | 9 - 10 | 1449 | 58 | |
More Information
Set Membership: |
|

SWAP70
Genomic Map