Assay Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098059.1 | NP_001091528.1 | 1 - 2 | 113 | 102 | |
| GenBank mRNA | BT030596.2 | - | 1 - 2 | 111 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001098059.1 transcripts.
Set Membership: |
|

USP12
Genomic Map