Assay Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098059.1 | NP_001091528.1 | 7 - 8 | 1006 | 128 | |
| GenBank mRNA | BC148869.1 | - | 2 - 3 | 451 | 128 | |
| BT030596.2 | - | 7 - 8 | 1004 | 128 | ||
More Information
Set Membership: |
|

USP12
Genomic Map