Assay Details
| Gene Symbol: SH3RF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098065.1 | NP_001091534.1 | 3 - 4 | 890 | 68 | |
| GenBank mRNA | BC140539.1 | - | 3 - 4 | 890 | 68 | |
More Information
Set Membership: |
|

SH3RF1
Genomic Map