Assay Details
| Gene Symbol: SERPINB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098080.1 | NP_001091549.1 | 6 - 7 | 766 | 76 | |
| GenBank mRNA | BC123811.1 | - | 6 - 7 | 766 | 76 | |
More Information
Set Membership: |
|

SERPINB5
Genomic Map