Assay Details
| Gene Symbol: SUFU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098083.2 | NP_001091552.1 | 3 - 4 | 710 | 67 | |
| GenBank mRNA | BC140543.1 | - | 3 - 4 | 712 | 67 | |
More Information
Set Membership: |
|

SUFU
Genomic Map