Assay Details
| Gene Symbol: YARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098088.2 | NP_001091557.1 | 2 - 3 | 1041 | 82 | |
| GenBank mRNA | BC140541.1 | - | 2 - 3 | 1042 | 82 | |
More Information
Set Membership: |
|

YARS2
Genomic Map