Assay Details
| Gene Symbol: SETD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098102.1 | NP_001091571.1 | 6 - 7 | 648 | 76 | |
| GenBank mRNA | BC140545.1 | - | 6 - 7 | 648 | 76 | |
More Information
Set Membership: |
|

SETD6
Genomic Map