Assay Details
| Gene Symbol: TINF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098114.1 | NP_001091583.1 | 9 - 10 | 2951 | 75 | |
| GenBank mRNA | BC133484.1 | - | 9 - 10 | 2951 | 75 | |
| BT030684.1 | - | 8 - 9 | 1793 | 75 | ||
More Information
Set Membership: |
|

TINF2
Genomic Map