Assay Details
| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098131.1 | NP_001091600.1 | 1 | 133 | 140 | |
| GenBank mRNA | BC114880.1 | - | 1 | 133 | 140 | |
More Information
Set Membership: |
|

RPL22L1
Genomic Map